2,3-Diethylpyrazine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:15707-24-1
:
Formula:C₈H₁₂N₂
MDL Number:MFCD00006151
:136.2 g/mol
Boiling Pt:180 °C (1013 hPa)
Density:0.97 g/cm³ (20 °C)
EINECS:239-800-1
UN:3334
ADR:9,
:M-Clarity™ quality level = MQ300