
2,6-Dichloropyrazine, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:4774-14-5
:
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00006125
:148.98 g/mol
Boiling Pt:120…122 °C (40 mmHg)
Melting Pt:57…58 °C
Density:1.713 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:225-316-8
:GHS07