2,6-Dichloropyrazine ≥98%
Supplier: Thermo Fisher Scientific
Jump to:
About this item
Purity: 99% CAS No.: 4774-14-5 Molecular Formula: C4H2Cl2N2 Formula Weight: 148.98 Flash Point: 110°(230°F) Melting Point: 52-56° Beilstein Reference: 2439 MDL No.: MFCD00006125
- CAS Number:4774-14-5
- Formula:C₄H₂Cl₂N₂
- MDL Number:MFCD00006125
- Molecular Weight:148.98 g/mol
- Boiling Pt:120…122 °C (40 mmHg)
- Melting Pt:52…56 °C
- Density:1.713 g/cm³ (20 °C)
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:225-316-8
3 Options Available Below
Product Details & Documents
Purity: 99% CAS No.: 4774-14-5 Molecular Formula: C4H2Cl2N2 Formula Weight: 148.98 Flash Point: 110°(230°F) Melting Point: 52-56° Beilstein Reference: 2439 MDL No.: MFCD00006125
- CAS Number:4774-14-5
- Formula:C₄H₂Cl₂N₂
- MDL Number:MFCD00006125
- Molecular Weight:148.98 g/mol
- Boiling Pt:120…122 °C (40 mmHg)
- Melting Pt:52…56 °C
- Density:1.713 g/cm³ (20 °C)
- Flash Pt:>110 °C (230 °F)
- Storage Temperature:Ambient
- EINECS:225-316-8



