
2,3-Dichloropyrazine 98%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:4858-85-9
:
Formula:C₄H₂Cl₂N₂
MDL Number:MFCD00040964
:148.98 g/mol
Boiling Pt:105…107 °C (50 mmHg)
Melting Pt:22.5…24.5 °C
Density:1.431 g/cm³
Flash Pt:102 °C
EINECS:225-460-1
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- GC:≥97.5 %