1-Methylxanthine
Glass bottle for solids, 100 mg
Catalog # MOLEM62653874
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: 3,7-Dihydro-1-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-1-methylpurine, 1-Methyl-1H-purine-2,6(3H,7H)-dione
- CAS Number:6136-37-4
- Formula:C₆H₆N₄O₂
- MDL Number:MFCD00005561
- Molecular Weight:166.14 g/mol
- EINECS:228-108-5
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg



