Order Entry
Export
Orders LinkContactUsLinkComponent
Guanine

Guanine

Glass bottle for solids, 100 g
Catalog # MOLE52634391-100G
Supplier:   Molekula
Guanine
Catalog # MOLE52634391-100G
Supplier:  Molekula
CAS Number:  73-40-5
Guanine

Glass bottle for solids, 100 g

CAS Number:  73-40-5
Price
125,00 €
Log in for availability
1 * 100 g
Product Details & Documents
Synonyms:  2-Amino-1H-purin-6(7H)-one
  • CAS Number:73-40-5
  • Formula:C₅H₅N₅O
  • MDL Number:MFCD00071533
  • Molecular Weight:151.13 g/mol
  • Merck Index:13,04580
  • Melting Pt:> 220 °C
  • Density:1.67 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:200-799-8
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Cat. No.:
    MOLE52634391-100G

Specifications