2,6-Dichloropurine
Catalog # MOLE62262074-1G
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
Specifications
About this item
:
:189 g/mol
Melting Pt:181 °C
EINECS:226-681-6
CAS Number:5451-40-1
Formula:C₅H₂Cl₂N₄
MDL Number:MFCD00077725