18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane), Sigma-Aldrich®
Catalog # SIAL274984-1G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
- Environmentally Preferable:
Specifications
About this item
Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6
:
:264.32 g/mol
Merck Index:13,02630
Boiling Pt:396 °C (1013 hPa)
Melting Pt:37…39 °C
Density:1.237 g/cm³ (25 °C)
:M-Clarity™ quality level = MQ200
Flash Pt:109 °C
Storage Temperature:Ambient
EINECS:241-473-5
:GHS07
CAS Number:17455-13-9
Formula:C₁₂H₂₄O₆
MDL Number:MFCD00005113