18-Crown-6 (1,4,7,10,13,16-hexaoxacyclooctadecane) 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
Synonyms: 1,4,7,10,13,16-Hexaoxacyclooctadecane, 18-Crown-6
CAS Number:17455-13-9
:
Formula:C₁₂H₂₄O₆
MDL Number:MFCD00005113
:264.32 g/mol
Merck Index:13,02630
Boiling Pt:396 °C (1013 hPa)
Melting Pt:36…40 °C
Density:1,237 g/cm³ (25 °C)
Flash Pt:>110 °C
EINECS:241-473-5
:GHS07
Storage Temperature:Ambient
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:Clear colorless to white Crystals or crystalline mass or liquid
- Infrared spectrum:Conforms
- Melting point:36 to 40 °C
- GC:≥98.5 %