Order Entry
Export
Orders LinkContactUsLinkComponent
 

Ethidium bromide, Sigma-Aldrich®

Toxic
Health
Supplier:   Merck
Ethidium bromide, Sigma-Aldrich®
Supplier:  Merck
Certificates
Ethidium bromide, Sigma-Aldrich®

4 Options Available Below

Product Details & Documents
Synonyms:  3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
  • CAS Number:1239-45-8
  • Formula:C₂₁H₂₀BrN₃
  • MDL Number:MFCD00011724
  • Molecular Weight:394.31 g/mol
  • Merck Index:13,04751
  • Boiling Pt:72 °C (0.1 mmHg)
  • Melting Pt:261…264 °C
  • Density:1.01 g/cm³ (20 °C)
  • Flash Pt:<100 °C
  • Storage Temperature:Ambient
  • UN:2811
  • ADR:6.1,I
  •  :M-Clarity™ quality level = MQ200
  • EINECS:214-984-6
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    1 g
    Log in for availability
    Log in for availability
    1 * 1 g
    1 * 1 g - 62,00 €
    62,00 €
    1 * 1 g
       62,00 €

  • Glass bottle for solids
    5 g
    Log in for availability
    Log in for availability
    1 * 5 g
    1 * 5 g - 244,00 €
    244,00 €
    1 * 5 g
       244,00 €

  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 421,00 €
    421,00 €
    1 * 10 g
       421,00 €

  • Glass bottle for solids
    25 g
    Log in for availability
    Log in for availability
    1 * 25 g
    1 * 25 g - 826,00 €
    826,00 €
    1 * 25 g
       826,00 €