Ethidium bromide, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
- CAS Number:1239-45-8
- Formula:C₂₁H₂₀BrN₃
- MDL Number:MFCD00011724
- Molecular Weight:394.31 g/mol
- Merck Index:13,04751
- Boiling Pt:72 °C (0.1 mmHg)
- Melting Pt:261…264 °C
- Density:1.01 g/cm³ (20 °C)
- Flash Pt:<100 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,I
- :M-Clarity™ quality level = MQ200
- EINECS:214-984-6
3 Options Available Below
Product Details & Documents
Synonyms: 3,8-Diamino-5-ethyl-6-phenylphenanthridinium bromide, 3,8-Diamino-5-ethyl-6-phenylphenanthridin-5-ium bromide
- CAS Number:1239-45-8
- Formula:C₂₁H₂₀BrN₃
- MDL Number:MFCD00011724
- Molecular Weight:394.31 g/mol
- Merck Index:13,04751
- Boiling Pt:72 °C (0.1 mmHg)
- Melting Pt:261…264 °C
- Density:1.01 g/cm³ (20 °C)
- Flash Pt:<100 °C
- Storage Temperature:Ambient
- UN:2811
- ADR:6.1,I
- :M-Clarity™ quality level = MQ200
- EINECS:214-984-6



