Perylene 99+%
- :198-55-0
- :C₂₀H₁₂
- :MFCD00004142
- :252.32 g/mol
- :12,07325
- :443 °C (1013 hPa)
- :277…280 °C
- :1,362 g/cm³ (20 °C)
- :Ambient
- :205-900-9
- :198-55-0
- :C₂₀H₁₂
- :MFCD00004142
- :252.32 g/mol
- :12,07325
- :443 °C (1013 hPa)
- :277…280 °C
- :1,362 g/cm³ (20 °C)
- :Ambient
- :205-900-9
- Appearance:Ochre to brown Powder
- Infrared spectrum:Conforms
- Melting point:277 to 280 °C
- HPLC:≥99.0 %
Product Family Options
- Pack typePk



