Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Sodium L(+)-ascorbate
Sodium L(+)-ascorbate
Sodium L(+)-ascorbate
EHERC10303900
:
EHRENSTORFER
:
Sodium L(+)-ascorbate
EHERC10303900
:
EHRENSTORFER
:
Pack type:
Glass bottle for solids
Pk:
0,25 g
Seq:
3029
:
:
198.11 g/mol
:
219 °C
:
1.799 g/cm³ (20 °C)
:
Ambient
:
205-126-1
:
134-03-2
:
C₆H₇NaO₆
:
MFCD00082340