0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Hexadecan-16-olide
Hexadecan-16-olide, Sigma-Aldrich®
Hexadecan-16-olide, Sigma-Aldrich®
Hexadecan-16-olide, Sigma-Aldrich®
Supplier:
Merck
Certificates
Expand image
Hexadecan-16-olide, Sigma-Aldrich®
Supplier:
Merck
About this item
CAS Number:
109-29-5
:
Formula:
C₁₆H₃₀O₂
MDL Number:
MFCD00039668
:
254.41 g/mol
:
M-Clarity™ quality level = MQ100
Storage Temperature:
Freezer
EINECS:
203-662-0