Ruthenium(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:12036-10-1
:
Formula:O₂Ru
MDL Number:MFCD00074857
Melting Pt:1200 °C
Density:6.97 g/cm³ (20 °C)
ADR:5.1,II
:M-Clarity™ quality level = MQ100
:133.07 g/mol
EINECS:234-840-6
UN:1479