
Ruthenium(III) chloride, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:10049-08-8
:
Formula:Cl₃Ru
MDL Number:MFCD00011208
:207.43 g/mol
Merck Index:12,08455
Melting Pt:500 °C
Density:3.11 g/cm³ (20 °C)
EINECS:233-167-5
ADR:8,III
:M-Clarity™ quality level = MQ200
UN:3260
:GHS05