


Platinum(IV) chloride 99%
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:13454-96-1
:
Formula:Cl₄Pt
MDL Number:MFCD00011182
Melting Pt:370 °C
Density:4,3 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:236-645-1
UN:2923
ADR:8,II
:336.89 g/mol
:GHS06
:GHS08
:GHS05
Specification Test Results
- Appearance:Brown Powder and/or chunks
- Trace metal analysis:≤500 ppm (Sum Ru+Rh+Ir+Pd)
- Calcium (Ca):≤250 ppm
- Copper (Cu):≤25 ppm
- Iron (Fe):≤50 ppm
- Lead (Pb):≤25 ppm
- Magnesium (Mg):≤250 ppm
- Platinum (Pt):57.0 to 58.5 %
- Silicon (Si):≤200 ppm