Order Entry
Export
Orders LinkContactUsLinkComponent
Iron(III) bromide, Sigma-Aldrich®

Iron(III) bromide, Sigma-Aldrich®

Irritant
Supplier:   Merck
Iron(III) bromide, Sigma-Aldrich®
Supplier:  Merck
Certificates
Iron(III) bromide, Sigma-Aldrich®

2 Options Available Below

Product Details & Documents
Synonyms:  Ferric bromide, Iron tribromide
  • CAS Number:10031-26-2
  • Formula:Br₃Fe
  • MDL Number:MFCD00016082
  • Molecular Weight:295.56 g/mol
  • Merck Index:12,04060
  • ADR:8,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:233-089-1
  • UN:3260
Product Family Options

Product Information

  • Pack type
    Pk
    Availability
    Price
  • Glass bottle for solids
    10 g
    Log in for availability
    Log in for availability
    1 * 10 g
    1 * 10 g - 93,30 €
    93,30 €
    1 * 10 g
       93,30 €

  • Glass bottle for solids
    50 g
    Log in for availability
    Log in for availability
    1 * 50 g
    1 * 50 g - 397,00 €
    397,00 €
    1 * 50 g
       397,00 €