Order Entry
Export
Orders LinkContactUsLinkComponent
Iron(III) bromide, anhydrous 98+%

Iron(III) bromide, anhydrous 98+%

Corrosive
Irritant
Iron(III) bromide, anhydrous 98+%
Certificates
Iron(III) bromide, anhydrous 98+%
Iron(III) bromide, anhydrous 98+%

3 Options Available Below

Product Details & Documents

Purity: 98+% CAS No.: 10031-26-2 Molecular Formula: FeBr3 Formula Weight: 295.57 Physical Form: Crystalline Sensitivity: Light Sensitive and Hygroscopic Appearance: Dark red Odour: Odourless Melting Point: dec. Merck Reference: 14,4018 MDL No.: MFCD00016082

Synonyms:  Ferric bromide, Iron tribromide
  • CAS Number:10031-26-2
  • Formula:Br₃Fe
  • MDL Number:MFCD00016082
  • Molecular Weight:295.56 g/mol
  • Merck Index:14,4018
  • Melting Pt:Dec.
  • Storage Temperature:Ambient
  • EINECS:233-089-1
  • UN:3260
  • ADR:8,III
Product Family Options

Product Information