

Manganese(IV) oxide
Catalog # ROTH4486.1
Supplier: Roth Carl
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 g
Specifications
About this item
:
:86.94 g/mol
Merck Index:14,05730
Melting Pt:535 °C
Density:5.03 g/cm³ (20 °C)
EINECS:215-202-6
:GHS03
:GHS07
CAS Number:1313-13-9
Formula:MnO₂
MDL Number:MFCD00003463