Tungsten trioxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1314-35-8
:
Formula:O₃W
MDL Number:MFCD00011466
:231.84 g/mol
Merck Index:12,09947
Melting Pt:1473 °C
Density:7.16 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ200
EINECS:215-231-4
:GHS07