Tungsten trioxide 99+%
Supplier: Thermo Fisher Scientific
About this item
Synonyms: Tungsten (VI) oxide
- CAS Number:1314-35-8
- Formula:O₃W
- MDL Number:MFCD00011466
- Molecular Weight:231.84 g/mol
- Merck Index:12,09947
- Melting Pt:1470…1475 °C
- Density:7,16 g/cm³ (20 °C)
- :Density: 7.16 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-231-4
2 Options Available Below
Product Details & Documents
Synonyms: Tungsten (VI) oxide
- CAS Number:1314-35-8
- Formula:O₃W
- MDL Number:MFCD00011466
- Molecular Weight:231.84 g/mol
- Merck Index:12,09947
- Melting Pt:1470…1475 °C
- Density:7,16 g/cm³ (20 °C)
- :Density: 7.16 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:215-231-4
Documents
ACRO21421
SDS
Specifications for Product Family
Specification Test Results
- Appearance:Yellow to yellow-green Powder
- Assay:≥99.0 %



