Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Niobium (V) oxide
Niobium(V) oxide
Niobium(V) oxide
Niobium(V) oxide
Supplier:
Roth Carl
Certificates
Expand image
Niobium(V) oxide
Supplier:
Roth Carl
About this item
CAS Number:
1313-96-8
:
Formula:
Nb₂O₅
MDL Number:
MFCD00011128
:
265.81 g/mol
Merck Index:
14,06559
Melting Pt:
1496 °C
Density:
4.6 g/cm³ (20 °C)
EINECS:
215-213-6