Niobium(V) oxide 99.99% (trace metals basis)
Supplier: Thermo Fisher Scientific
Certificates
About this item
CAS Number:1313-96-8
:
Formula:Nb₂O₅
MDL Number:MFCD00011128
:265.81 g/mol
Merck Index:14,06559
Melting Pt:1520 °C
Density:4.47 g/cm³
Storage Temperature:Ambient
EINECS:215-213-6
Specification Test Results
- Appearance:White to off-white Powder
- Total trace metal impurities:≤0.01 %