Titanium(IV) ethanolate, Sigma-Aldrich®
Catalog # SIAL86710-50ML
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:50 mL
- Seq:2370
Specifications
About this item
:
:228.12 g/mol
Boiling Pt:150 °C (1013 hPa)
Density:1.107 g/cm³ (20 °C)
Flash Pt:33 °C
EINECS:221-410-8
UN:2924
ADR:3,II
CAS Number:3087-36-3
Formula:C₈H₂₀O₄Ti
MDL Number:MFCD00009071