Titanium(IV) ethanolate, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:3087-36-3
:
Formula:C₈H₂₀O₄Ti
MDL Number:MFCD00009071
:228.12 g/mol
Boiling Pt:150 °C (1013 hPa)
Density:1.107 g/cm³ (20 °C)
Flash Pt:33 °C
EINECS:221-410-8
ADR:3,II
:M-Clarity™ quality level = MQ100
UN:2924