
Zirconium(IV) oxide, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:1314-23-4
:
Formula:ZrO₂
MDL Number:MFCD00011310
:123.22 g/mol
Merck Index:14,10180
Boiling Pt:4300 °C (1013 hPa)
Melting Pt:2680 °C
Density:5.75 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
EINECS:215-227-2
:GHS07
Storage Temperature:Ambient