Base plate, MicroAmp™, 96-well
Catalog # APBI4312063
Supplier: APPLIED BIOSYSTEMS
Specifications
- Description:Base plate, 96-well microplate
- Pk:10
Specifications
About this item
MicroAmp® bases are designed to hold reaction tubes in tray or retainer sets or 96-well reaction plates for convenient pre-PCR and post-PCR sample processing.
- Ideal for centrifugation
Use with instruments that have heated sample covers to eliminate condensation and the need for a mineral oil overlay. MicroAmp® reaction tubes are essential for high performance PCR. They ensure the greatest temperature and amplification uniformity for your samples.