ContactUsLinkComponent
Titanium(IV) oxide, Sigma-Aldrich®
  SIAL635057-1EA
 :  Merck
 :  
Titanium(IV) oxide, Sigma-Aldrich®
  SIAL635057-1EA
 :  Merck
 :  
  • Pk:
    1
  • Seq:
    0001
 :
 :79.88 g/mol
 :13,09549
 :2900 °C (1013 hPa)
 :1855 °C
 :4.2 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
 :Ambient
 :236-675-5
 :13463-67-7
 :1317-70-0
 :TiO₂
 :MFCD00011269