Order Entry
Export
ContactUsLinkComponent
Zinc sulphide, Sigma-Aldrich®
Zinc sulphide, Sigma-Aldrich®
Catalog # SIAL333271-50G
Supplier:  Merck
CAS Number:  
Zinc sulphide, Sigma-Aldrich®
Catalog # SIAL333271-50G
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368

Specifications

About this item

 :
 :97.46 g/mol
Merck Index:12,10294
Melting Pt:1064 °C
Density:4.1 g/cm³ (20 °C)
 :M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:215-251-3
CAS Number:1314-98-3
Formula:SZn
MDL Number:MFCD00011301