0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Zinc sulphide
Zinc sulphide ≥99.99% (metals basis)
Zinc sulphide ≥99.99% (metals basis)
:
Thermo Fisher Scientific
Zinc sulphide ≥99.99% (metals basis)
:
Thermo Fisher Scientific
:
1314-98-3
:
:
SZn
:
MFCD00011301
:
97.46 g/mol
:
14,10160
:
1700 °C
:
4.10
:
Ambient
:
215-251-3
msds_ALFA040091
Thermo Scientific Acroseal 25 Years