Zinc oxide, Sigma-Aldrich®
Supplier: Merck
Jump to:
About this item
Synonyms: Zinci oxidum, Zinc (II) oxide
- CAS Number:1314-13-2
- Formula:ZnO
- MDL Number:MFCD00011300
- Molecular Weight:81.39 g/mol
- Merck Index:13,10200
- Melting Pt:1975 °C
- Density:5.61 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:215-222-5
- UN:3077
- ADR:9,III
- :M-Clarity™ quality level = MQ200
- REACH:01-2119463881-32
2 Options Available Below
Product Details & Documents
Synonyms: Zinci oxidum, Zinc (II) oxide
- CAS Number:1314-13-2
- Formula:ZnO
- MDL Number:MFCD00011300
- Molecular Weight:81.39 g/mol
- Merck Index:13,10200
- Melting Pt:1975 °C
- Density:5.61 g/cm³ (25 °C)
- Storage Temperature:Ambient
- EINECS:215-222-5
- UN:3077
- ADR:9,III
- :M-Clarity™ quality level = MQ200
- REACH:01-2119463881-32



