ContactUsLinkComponent
Mercury (II) oxide, red
Mercury (II) oxide, red
  ROTH4470.1
 :  Roth Carl
 :  
Mercury (II) oxide, red
  ROTH4470.1
 :  Roth Carl
 :  
  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g
  • Seq:
    3368
 :
 :216.59 g/mol
 :13,05908
 :356.6 °C (1013 hPa)
 :> 400 °C
 :11 g/cm³ (20 °C)
 :Ambient
 :244-654-7
 :1641
 :6.1,II
 :GHS06
 :GHS08
 :GHS09
 :21908-53-2
 :HgO
 :MFCD00011045