Zinc fluoride 98%
Supplier: Thermo Fisher Scientific
About this item
- CAS Number:7783-49-5
- Formula:F₂Zn
- MDL Number:MFCD00011298
- Molecular Weight:103.39 g/mol
- Boiling Pt:1500 °C (1013 hPa)
- Melting Pt:872…910 °C
- Density:4,95 g/cm³ (20 °C)
- EINECS:232-001-9
- ADR:6.1,III
- :Density: 4.95 g/cm³ (20 °C)
2 Options Available Below
Product Details & Documents
- CAS Number:7783-49-5
- Formula:F₂Zn
- MDL Number:MFCD00011298
- Molecular Weight:103.39 g/mol
- Boiling Pt:1500 °C (1013 hPa)
- Melting Pt:872…910 °C
- Density:4,95 g/cm³ (20 °C)
- EINECS:232-001-9
- ADR:6.1,III
- :Density: 4.95 g/cm³ (20 °C)
Documents
ACRO36391
SDS
spec_ACRO36391.pdf
Product specification
Specifications for Product Family
Specification Test Results
- Appearance:White to off-white Powder
- Fluoride (F):36.0 to 37.8 %



