
Acenaphthene
Catalog # EHERC20505000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 mg
- Seq:2440
Specifications
About this item
:
:154.21 g/mol
Merck Index:13,00029
Boiling Pt:279 °C (1013 hPa)
Melting Pt:90…93 °C
Density:1.024 g/cm³ (20 °C)
Flash Pt:135 °C (open cup)
Storage Temperature:Ambient
EINECS:201-469-6
UN:3077
ADR:9,III
:GHS09
CAS Number:83-32-9
Formula:C₁₂H₁₀
MDL Number:MFCD00003807