2,2'-Binaphthyl
Catalog # EHERC20660000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
:254.33 g/mol
Boiling Pt:452 °C (1013 hPa)
Boiling Pt:452 °C (1013 hPa)
Melting Pt:188.3 °C
Density:1.14 g/cm³ (20 °C)
EINECS:210-320-4
CAS Number:612-78-2
Formula:C₂₀H₁₄
MDL Number:MFCD00021584