
1,1'-Binaphthyl
Catalog # EHERC20655000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:10 mg
- Seq:2275
Specifications
About this item
:
Boiling Pt:405 °C (1013 hPa)
Melting Pt:142…144 °C
Density:1.19 g/cm³ (20 °C)
:254.33 g/mol
EINECS:210-070-6
:GHS07
CAS Number:604-53-5
Formula:C₂₀H₁₄
MDL Number:MFCD00041740