2,6-Dimethylnaphthalene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:581-42-0
:
Formula:C₁₂H₁₂
MDL Number:MFCD00004120
:156.23 g/mol
Boiling Pt:169.8 °C (1013 hPa)
Melting Pt:–44.7 °C
Density:1.8682 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-464-0