2,3-Dimethylnaphthalene, Sigma-Aldrich®
SIALD170801-1G
: Merck
:
- Pack type:Glass bottle for solids
- Pk:1 g
- Seq:3047
:
:156.23 g/mol
:269 °C (1013 hPa)
:103…104 °C
:1.003 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:Ambient
:209-463-5
:GHS07
:581-40-8
:C₁₂H₁₂
:MFCD00004119