1,6-Dimethylnaphthalene, Sigma-Aldrich®
Catalog # SIAL250937-250MG
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:250 mg
- Seq:2590
Specifications
About this item
:
Boiling Pt:265 °C (1013 hPa)
Melting Pt:–16.9 °C
Density:1.009 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:156.23 g/mol
Storage Temperature:Ambient
EINECS:209-385-1
CAS Number:575-43-9
Formula:C₁₂H₁₂
MDL Number:MFCD00004039