
1,5-Dimethylnaphthalene
Catalog # EHERC20762000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:50 mg
- Seq:2370
Specifications
About this item
:
:156.23 g/mol
Boiling Pt:265…266 °C (1013 hPa)
Melting Pt:75 °C
EINECS:209-338-5
:GHS07
CAS Number:571-61-9
Formula:C₁₂H₁₂
MDL Number:MFCD00004038