1,3-Dimethylnaphthalene, Sigma-Aldrich®
Catalog # SIALD170208-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
Boiling Pt:263 °C (1013 hPa)
Melting Pt:–4 °C
Density:1.0146 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
:156.23 g/mol
Storage Temperature:Ambient
EINECS:209-384-6
CAS Number:575-41-7
Formula:C₁₂H₁₂
MDL Number:MFCD00004036