Order Entry
Export
Orders LinkContactUsLinkComponent
 

2,7-Dihydroxynaphthalene 97%

2,7-Dihydroxynaphthalene 97%
Certificates
2,7-Dihydroxynaphthalene 97%

4 Options Available Below

Product Details & Documents
Synonyms:  2,7-Naphthalenediol, Naphthalene-2,7-diol
  • CAS Number:582-17-2
  • Formula:C₁₀H₈O₂
  • MDL Number:MFCD00004085
  • Molecular Weight:160.17 g/mol
  • Boiling Pt:375 °C (1013 hPa)
  • Melting Pt:184…189 °C
  • Storage Temperature:Ambient
  • EINECS:209-478-7
Specifications for Product Family

Specification Test Results

  • Appearance:
    Grey Powder
  • Infrared spectrum:
    Conforms
  • Melting point:
    184 to 189 °C
  • GC:
    ≥96.0 %
Product Family Options

Product Information