
2,6-Dihydroxynaphthalene, Sigma-Aldrich®
Catalog # SIAL275603-5G
Supplier: Merck
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g
- Seq:3173
Specifications
About this item
:
:160.17 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:220 °C
Density:1.33 g/cm³ (20 °C)
:M-Clarity™ quality level = MQ100
Storage Temperature:Ambient
EINECS:209-465-6
:GHS07
CAS Number:581-43-1
Formula:C₁₀H₈O₂
MDL Number:MFCD00004082