2,6-Dihydroxynaphthalene
Glass bottle for solids, 5 g
Catalog # MOLEM45946669
Supplier: Molekula
Jump to:
Product Details & Documents
Synonyms: Naphthalene-2,6-diol
- CAS Number:581-43-1
- Formula:C₁₀H₈O₂
- MDL Number:MFCD00004082
- Molecular Weight:160.17 g/mol
- Boiling Pt:375 °C (1013 hPa)
- Melting Pt:220 °C
- Density:1.33 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:209-465-6
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



