Order Entry
Export
Orders LinkContactUsLinkComponent
2,6-Dihydroxynaphthalene ≥98%

2,6-Dihydroxynaphthalene ≥98%

2,6-Dihydroxynaphthalene ≥98%
Certificates
2,6-Dihydroxynaphthalene ≥98%
2,6-Dihydroxynaphthalene ≥98%

3 Options Available Below

Product Details & Documents

Purity: 98% CAS No.: 581-43-1 Molecular Formula: C10H8O2 Formula Weight: 160.17 Melting Point: 223-225° Beilstein Reference: 1238082

Synonyms:  Naphthalene-2,6-diol
  • CAS Number:581-43-1
  • Formula:C₁₀H₈O₂
  • MDL Number:MFCD00004082
  • Molecular Weight:160.17 g/mol
  • Boiling Pt:375 °C (1013 hPa)
  • Melting Pt:223…225 °C
  • Density:1.33 g/cm³ (20 °C)
  • Storage Temperature:Ambient
  • EINECS:209-465-6
Product Family Options

Product Information