2,3-Dihydroxynaphthalene, Sigma-Aldrich®
Supplier: Merck
Certificates
About this item
CAS Number:92-44-4
:
Formula:C₁₀H₈O₂
MDL Number:MFCD00004073
:160.17 g/mol
Boiling Pt:354 °C (1013 hPa)
Melting Pt:151…152 °C
Density:1.12 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:202-156-7