
1,6-Dihydroxynaphthalene
Catalog # MOLEM77000905
Supplier: Molekula
CAS Number:
Specifications
- Pack type:Glass bottle for solids
- Pk:100 g
- Seq:3401
Specifications
About this item
:
:160.17 g/mol
Boiling Pt:375 °C (1013 hPa)
Melting Pt:135.5 °C
EINECS:209-386-7
:GHS07
Storage Temperature:Ambient
CAS Number:575-44-0
Formula:C₁₀H₈O₂
MDL Number:MFCD00003981
Boiling Pt:375 °C (1013 hPa)