Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
1,5-Dihydroxynaphthalene
1,5-Dihydroxynaphthalene ≥98%
1,5-Dihydroxynaphthalene ≥98%
:
Thermo Fisher Scientific
1,5-Dihydroxynaphthalene ≥98%
:
Thermo Fisher Scientific
:
83-56-7
:
:
C₁₀H₈O₂
:
MFCD00003980
:
160.17 g/mol
:
375 °C (1013 hPa)
:
∼ 260 °C (dec.)
:
1.41 g/cm³ (20 °C)
:
252 °C (486 °F)
:
Ambient
:
201-487-4
:
3077
:
9,III
:
GHS08
:
GHS07
msds_ALFAL04599
Organic Building Blocks and Intermediates
Thermo Scientific Acroseal 25 Years