Order Entry
Export
ContactUsLinkComponent
1,4-Dihydroxynaphthalene
1,4-Dihydroxynaphthalene
Catalog # MOLE61355361-5G
Supplier:  Molekula
CAS Number:  
1,4-Dihydroxynaphthalene
Catalog # MOLE61355361-5G
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    5 g
  • Seq:
    3173

Specifications

About this item

 :
 :160.17 g/mol
Melting Pt:191…193 °C
Density:1,1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:209-336-4
CAS Number:571-60-8
Formula:C₁₀H₈O₂
MDL Number:MFCD00003977